4-Chlorobenzoylformic acid 95%

Supplier: Ambeed
Warning

Synonyms: 2-(4-Chlorophenyl)-2-oxoacetic acid

A216680-250MG A216680-1G
103818-572EA 108 USD
103818-572 103849-294 76873-820 77087-886
4-Chlorobenzoylformic acid 95%
4-Chlorobenzoylformic acid

Precaución: for Research Use Only

Formula: C₈H₅ClO₃
MW: 184.577 g/mol
Boiling Pt: 309.0±34.0 °C (760 mm Hg)
Flash Pt: 140.7 °C
Storage Temperature: Ambient
MDL Number: MFCD02656397
CAS Number: 7099-88-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR