4-Chloro-3-fluorophenethyl alcohol 95%

Supplier: Ambeed
Warning

Synonyms: 2-(4-Chloro-3-fluorophenyl)ethanol

A808808-1G A808808-5G A808808-10G
103841-084EA 314 USD
103841-084 76845-336 76845-338 77159-266
4-Chloro-3-fluorophenethyl alcohol 95%
4-Chloro-3-fluorophenethyl alcohol

Precaución: for Research Use Only

Formula: C₈H₈ClFO
Storage Temperature: Ambient
MDL Number: MFCD06201151
CAS Number: 206362-79-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR