(4-Chloro-3-fluorophenyl)acetic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-(4-Chloro-3-fluorophenyl)acetic acid

A249500-25G A249500-5G A249500-10G A249500-250MG
103811-878EA 11 USD
103811-878 103851-716 77091-054 77091-056 77288-970 77288-972
(4-Chloro-3-fluorophenyl)acetic acid 98%
(4-Chloro-3-fluorophenyl)acetic acid

Precaución: for Research Use Only

Formula: (C₆H₃ClF)CH₂COOH
MW: 188.59 g/mol
Melting Pt: 88…91 °C
Storage Temperature: Ambient
MDL Number: MFCD04115861
CAS Number: 883500-51-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR