2-(4-Bromopyridin-2-yl)acetonitrile 97%

Supplier: Ambeed
Warning

A442359-1G A442359-100MG A442359-10G A442359-250MG A442359-5G
103827-684EA 101 USD
103827-684 103838-754 76842-524 76990-620 76990-622 76990-624 77307-318
2-(4-Bromopyridin-2-yl)acetonitrile 97%
2-(4-Bromopyridin-2-yl)acetonitrile

Precaución: for Research Use Only

Formula: C₇H₅BrN₂
Boiling Pt: 301.582 °C (760 mm Hg)
Flash Pt: 136.192 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09999985
CAS Number: 312325-73-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR