2-(4-Bromophenyl)thiophene 97%

Supplier: Ambeed
Warning

A361365-1G A361365-5G A361365-250MG
103826-280EA 45 USD
103826-280 103837-294 77306-722 77306-724 77626-696
2-(4-Bromophenyl)thiophene 97%
2-(4-Bromophenyl)thiophene

Precaución: for Research Use Only

Formula: C₁₀H₇BrS
MW: 239.13 g/mol
Boiling Pt: 307 °C (760 mm Hg)
Melting Pt: 95…99 °C
Storage Temperature: Refrigerator
MDL Number: MFCD05664340
CAS Number: 40133-22-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR