2-(4-Bromophenyl)pyrrolidine 97%

Supplier: Ambeed
Warning

A265918-250MG A265918-1G
103812-766EA 118 USD
103812-766 103852-662 76977-120 77092-360
2-(4-Bromophenyl)pyrrolidine 97%
2-(4-Bromophenyl)pyrrolidine

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrN
MW: 226.1130 g/mol
Boiling Pt: 287.2 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD02663432
CAS Number: 383127-22-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR