2-(4-Bromophenyl)pyridine 98%

Supplier: Ambeed
Warning

A225187-1G A225187-25G A225187-5G A225187-10G
103819-164EA 6 USD
103819-164 103850-056 77088-734 77088-736 77088-738 77237-712
2-(4-Bromophenyl)pyridine 98%
2-(4-Bromophenyl)pyridine

Precaución: for Research Use Only

Formula: C₁₁H₈BrN
MW: 234.10 g/mol
Boiling Pt: 322.3 °C (760 mm Hg)
Flash Pt: 148.7 °C
Storage Temperature: Ambient
MDL Number: MFCD04116231
CAS Number: 63996-36-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR