2-(4-Bromophenyl)propan-2-ol 97%
Supplier: Ambeed
|
Warning
|
A512869-5G
A512869-250MG
103828-422EA
14
USD
103828-422
103839-520
76843-508
77882-261
2-(4-Bromophenyl)propan-2-ol 97%
2-(4-Bromophenyl)propan-2-ol
Precaución: for Research Use Only
Formula:
C₉H₁₁BrO Boiling Pt: 281.5 °C (760 mm Hg) Melting Pt: 45.6 °C Flash Pt: 124 °C Storage Temperature: Ambient |
MDL Number:
MFCD11870096 CAS Number: 2077-19-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...