2-(4-Bromophenyl)naphthalene 98% (by GC)

Supplier: Ambeed
Warning

A408967-1G A408967-25G A408967-5G A408967-100G A408967-250MG
103827-192EA 11 USD
103827-192 103838-240 76841-752 76841-754 76841-756 77240-720 77240-722
2-(4-Bromophenyl)naphthalene 98% (by GC)
2-(4-Bromophenyl)naphthalene

Precaución: for Research Use Only

Formula: C₁₆H₁₁Br
MW: 283.17 g/mol
Boiling Pt: 394.3 °C (760 mm Hg)
Melting Pt: 131…135 °C (760 mm Hg)
Flash Pt: 190.4 °C
Storage Temperature: Ambient
MDL Number: MFCD09271778
CAS Number: 22082-99-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR