2-(4-Bromophenyl)ethanol 98%

Supplier: Ambeed
Warning

Synonyms: 4-Bromophenethyl alcohol

A232809-100G A232809-10G A232809-1G A232809-500G A232809-25G A232809-5G
103850-606EA 8 USD
103850-606 76976-208 76976-210 76976-212 76976-214 77089-540 77089-542
2-(4-Bromophenyl)ethanol 98%
2-(4-Bromophenyl)ethanol

Precaución: for Research Use Only

Formula: C₈H₉BrO
MW: 201.06 g/mol
Boiling Pt: 274.6 °C (760 mm Hg)
Melting Pt: 36…38 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00002897
CAS Number: 4654-39-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR