2-(4-bromophenyl)cyclopropane-1-carboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: 2-(4-Bromophenyl)cyclopropanecarboxylic acid

A339803-100MG A339803-250MG
103825-800EA 354 USD
103825-800 103836-774 76828-462 76828-464
2-(4-bromophenyl)cyclopropane-1-carboxylic acid 95%
2-(4-bromophenyl)cyclopropane-1-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₉BrO₂
MW: 241.081 g/mol
Boiling Pt: 364.5±42.0 °C (760 mm Hg)
Melting Pt: 122 °C
Storage Temperature: Ambient
MDL Number: MFCD03196280
CAS Number: 77255-26-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR