2-(4-Bromophenyl)-4,6-diphenyl-1,3,5-triazine 98%

Supplier: Ambeed
Warning

A313100-100G A313100-10G A313100-250MG A313100-1G A313100-25G A313100-5G
103814-716EA 6 USD
103814-716 103835-906 76828-040 76828-042 76828-044 76838-724 76838-726 76838-728
2-(4-Bromophenyl)-4,6-diphenyl-1,3,5-triazine 98%
2-(4-Bromophenyl)-4,6-diphenyl-1,3,5-triazine

Precaución: for Research Use Only

Formula: C₂₁H₁₄BrN₃
MW: 388.27 g/mol
Melting Pt: 193…197 °C
Storage Temperature: Ambient
MDL Number: MFCD00194632
CAS Number: 23449-08-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR