N-(4-Bromobutyl)phthalimide 97%

Supplier: Ambeed
Warning

Synonyms: 2-(4-Bromobutyl)isoindoline-1,3-dione

A271735-100G A271735-10G A271735-500G A271735-25G A271735-1G A271735-5G
103813-062EA 5 USD
103813-062 103852-974 76723-040 76723-042 76723-044 77092-758 77305-096 77305-098
N-(4-Bromobutyl)phthalimide 97%
N-(4-Bromobutyl)phthalimide

Precaución: for Research Use Only

Formula: C₁₂H₁₂BrNO₂
MW: 282.14 g/mol
Boiling Pt: 381.3 °C (760 mm Hg)
Melting Pt: 76…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00005905
CAS Number: 5394-18-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR