(4-Bromo-3-methylphenyl)acetic acid 98%
Supplier: Ambeed
|
Warning
|
A100889-10G
A100889-250MG
A100889-1G
A100889-5G
103795-676EA
33
USD
103795-676
103821-366
77025-156
77025-158
77296-884
77296-886
(4-Bromo-3-methylphenyl)acetic acid 98%
(4-Bromo-3-methylphenyl)acetic acid
Precaución: for Research Use Only
Formula:
C₉H₉BrO₂ Boiling Pt: 338 °C (760 mm Hg) Melting Pt: 81 °C Flash Pt: 158 °C Storage Temperature: Ambient |
MDL Number:
MFCD02664811 CAS Number: 215949-57-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...