2-(4-Bromo-2-methoxyphenyl)acetic acid 97%
Supplier: Ambeed
|
Warning
|
A142345-100G
A142345-10G
A142345-250MG
A142345-1G
A142345-25G
A142345-5G
103803-146EA
17
USD
103803-146
76811-820
76811-822
76811-824
77316-366
77316-368
77316-370
2-(4-Bromo-2-methoxyphenyl)acetic acid 97%
2-(4-Bromo-2-methoxyphenyl)acetic acid
Precaución: for Research Use Only
Formula:
C₉H₉BrO₃ MW: 245.07 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11847505 CAS Number: 1026089-09-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...