(4-Bromo-2-fluorophenyl)acetonitrile 98%

Supplier: Ambeed
Warning

A225345-1G A225345-25G A225345-5G A225345-100G A225345-10G A225345-500G
103819-174EA 6 USD
103819-174 103850-068 77088-756 77088-758 77088-760 77288-282 77288-284 77288-286
(4-Bromo-2-fluorophenyl)acetonitrile 98%
(4-Bromo-2-fluorophenyl)acetonitrile

Precaución: for Research Use Only

Formula: C₈H₅BrFN
Boiling Pt: 281.2 °C (760 mm Hg)
Melting Pt: 52.8…53 °C
Flash Pt: 123.8 °C
Storage Temperature: Ambient
MDL Number: MFCD07368653
CAS Number: 114897-91-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR