(4-Bromo-1H-pyrazol-1-yl)acetic acid 97%

Supplier: Ambeed
Warning

A218317-1G A218317-250MG
103818-706EA 37 USD
103818-706 103849-580 77303-484 77881-540
(4-Bromo-1H-pyrazol-1-yl)acetic acid 97%
(4-Bromo-1H-pyrazol-1-yl)acetic acid

Precaución: for Research Use Only

Formula: C₅H₅BrN₂O₂
Boiling Pt: 363.1 °C (760 mm Hg)
Flash Pt: 173.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00159632
CAS Number: 82231-53-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR