(4-Aminophenyl)acetonitrile 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
2-(4-Aminophenyl)acetonitrile, 4-Aminobenzyl cyanide
A913458-25G
A913458-5G
A913458-10G
103841-384EA
7
USD
103841-384
76845-658
76845-660
77256-588
(4-Aminophenyl)acetonitrile 98%
(4-Aminophenyl)acetonitrile
Precaución: for Research Use Only
Formula:
C₈H₈N₂ MW: 132.17 g/mol Boiling Pt: 312 °C (760 mm Hg) Melting Pt: 45…48 °C Flash Pt: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007912 CAS Number: 3544-25-0 UN: 3439 ADR: 6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...