2-(4-Aminophenyl)-6-methylbenzothiazole 95%

Supplier: Ambeed
Warning

Synonyms: 4-(6-methyl-1,3-benzothiazol-2-yl)aniline

A392919-1G A392919-25G A392919-5G A392919-100G A392919-10G A392919-250MG
103826-914EA 6 USD
103826-914 103837-954 76841-350 76841-352 76841-354 76876-804 76876-806 76876-808
2-(4-Aminophenyl)-6-methylbenzothiazole 95%
2-(4-Aminophenyl)-6-methylbenzothiazole

Precaución: for Research Use Only

Formula: C₁₄H₁₂N₂S
MW: 240.324 g/mol
Boiling Pt: 434 °C (760 mm Hg)
Melting Pt: 191…196 °C
Flash Pt: 217.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00005780
CAS Number: 92-36-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR