2-(4-Aminophenoxy)-N,N,N-trimethylethanaminium bromide hydrobromide 97%

Supplier: Ambeed
Warning

Synonyms: 4-APC HBr

A105668-250MG A105668-1G
103796-922EA 425 USD
103796-922 77160-470 77283-376
2-(4-Aminophenoxy)-N,N,N-trimethylethanaminium bromide hydrobromide 97%
2-(4-Aminophenoxy)-N,N,N-trimethylethanaminium bromide hydrobromide

Precaución: for Research Use Only

Formula: C₁₁H₂₀Br₂N₂O
MW: 356.1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22580414
CAS Number: 1076196-38-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR