2-(4-Amino-3-methylphenyl)-1,1,1,3,3,3-hexafluoropropan-2-ol 98%

Supplier: Ambeed
Warning

Synonyms: 4-(1,1,1,3,3,3-Hexafluoro-2-hydroxyprop-2-yl)-2-methylaniline, 2-Amino-5-(1,1,1,3,3,3-hexafluoro-2-hydroxyprop-2-yl)toluene

A130078-100MG A130078-250MG A130078-1G
103801-374EA 279 USD
103801-374 77009-874 77009-876 77313-806
2-(4-Amino-3-methylphenyl)-1,1,1,3,3,3-hexafluoropropan-2-ol 98%
2-(4-Amino-3-methylphenyl)-1,1,1,3,3,3-hexafluoropropan-2-ol

Precaución: for Research Use Only

Formula: C₁₀H₉F₆NO
MW: 273.175 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00788428
CAS Number: 1992-09-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR