2-(4-Amino-1-(tert-butoxycarbonyl)piperidin-4-yl)acetic acid 95%
Supplier: Ambeed
|
Warning
|
A167462-100MG
A167462-250MG
A167462-1G
103806-190EA
207
USD
103806-190
103844-452
76813-066
76813-068
77300-900
2-(4-Amino-1-(tert-butoxycarbonyl)piperidin-4-yl)acetic acid 95%
2-(4-Amino-1-(tert-butoxycarbonyl)piperidin-4-yl)acetic acid
Precaución: for Research Use Only
Formula:
C₁₂H₂₂N₂O₄ MW: 258.31 g/mol Boiling Pt: 398.9±27.0 °C (760 mm Hg) Flash Pt: 195.1 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD08460439 CAS Number: 1159983-30-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...