2-(4,6-Dichloropyrimidin-5-yl)acetaldehyde 95%

Supplier: Ambeed
Warning

Synonyms: 5-Acetaldehydeyl-4,6-dichloropyrimidine

A161316-250MG A161316-5G A161316-10G A161316-25G
103805-474EA 11 USD
103805-474 103843-712 76768-568 77319-680 77550-022 77567-352
2-(4,6-Dichloropyrimidin-5-yl)acetaldehyde 95%
2-(4,6-Dichloropyrimidin-5-yl)acetaldehyde

Precaución: For research use only.

Formula: C₆H₄Cl₂N₂O
MW: 191.0200 g/mol
Boiling Pt: 312.7 °C (760 mm Hg)
Melting Pt: 88…91 °C
Storage Temperature: Freezer
MDL Number: MFCD09991563
CAS Number: 16019-33-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR