4,4-Dimethylcyclohexene-1-ylboronic acid pinacol ester 95%

Supplier: Ambeed
Warning

Synonyms: 2-(4,4-Dimethyl-1-cyclohexen-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

A168808-100MG A168808-250MG A168808-1G A168808-5G
103806-320EA 24 USD
103806-320 103844-574 76813-128 76813-130 77300-946 77300-948
4,4-Dimethylcyclohexene-1-ylboronic acid pinacol ester 95%
4,4-Dimethylcyclohexene-1-ylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₂₅BO₂
Boiling Pt: 255 °C (760 mm Hg)
Flash Pt: 108 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08690233
CAS Number: 859217-67-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR