2-Hydroxyphenylboronic acid pinacol ester 95%

Supplier: Ambeed
Warning

Synonyms: 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenol

A124654-100G A124654-10G A124654-250MG A124654-1G A124654-5G A124654-25G
103800-564EA 8 USD
103800-564 76718-690 76718-692 76718-694 77312-702 77312-704 77549-476
2-Hydroxyphenylboronic acid pinacol ester 95%
2-Hydroxyphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₇BO₃
MW: 220.08 g/mol
Boiling Pt: 320.9 °C (760 mm Hg)
Melting Pt: 185 °C
Flash Pt: 147.9 °C
Storage Temperature: Ambient
MDL Number: MFCD02093754
CAS Number: 269409-97-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR