2-Carboxyphenylboronic acid, pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid

A166016-100G A166016-10G A166016-1G A166016-25G A166016-5G A166016-250MG
103806-012EA 11 USD
103806-012 103844-268 77151-082 77151-084 77320-494 77320-496 77320-498 77685-351
2-Carboxyphenylboronic acid, pinacol ester 97%
2-Carboxyphenylboronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₇BO₄
MW: 248.083 g/mol
Boiling Pt: 383 °C (760 mm Hg)
Flash Pt: 185.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08458199
CAS Number: 1187591-17-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR