2-Formylbenzeneboronic acid pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: 2-Formylphenylboronic acid Pinacol ester, 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde, 2-Formylphenylboronic acid pinacol ester, 2-(2-Formylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

A117333-100G A117333-10G A117333-25G A117333-1G A117333-5G
103798-866EA 11 USD
103798-866 103824-746 76718-368 76718-370 76718-372 77311-526 77311-528
2-Formylbenzeneboronic acid pinacol ester 97%
2-Formylbenzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₇BO₃
Boiling Pt: 345.9 °C (760 mm Hg)
Flash Pt: 163 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07363841
CAS Number: 380151-85-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR