2-Aminophenylboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 2-Aminobenzeneboronic acid pinacol ester

A187297-100G A187297-10G A187297-250MG A187297-500G A187297-1G A187297-5G A187297-25G
103816-002EA 14 USD
103816-002 103846-592 76721-044 76721-046 76721-048 76721-050 77084-278 77084-280 77302-076
2-Aminophenylboronic acid pinacol ester 98%
2-Aminophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₈BNO₂
MW: 219.09 g/mol
Boiling Pt: 335.6 °C (760 mm Hg)
Melting Pt: 63…68 °C
Flash Pt: 156.8 °C
Storage Temperature: Ambient
MDL Number: MFCD02179448
CAS Number: 191171-55-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR