2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-4-(trifluoromethyl)benzaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: 2-Formyl-5-(trifluoromethyl)phenylboronic acid pinacol ester

A111294-100MG A111294-250MG
103797-812EA 90 USD
103797-812 103823-626 77160-752 77160-754
2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-4-(trifluoromethyl)benzaldehyde 97%
2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-4-(trifluoromethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₁₆BF₃O₃
MW: 300.08 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18757662
CAS Number: 1416721-24-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR