4-(Trifluoromethyl)phenethyl alcohol 97%

Supplier: Ambeed
Warning

Synonyms: 2-(4-(Trifluoromethyl)phenyl)ethanol

A179208-1G A179208-250MG
103807-408EA 17 USD
103807-408 103845-754 77083-212 77287-104
4-(Trifluoromethyl)phenethyl alcohol 97%
4-(Trifluoromethyl)phenethyl alcohol

Precaución: for Research Use Only

Formula: C₉H₉F₃O
Boiling Pt: 218.2 °C (760 mm Hg)
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06411331
CAS Number: 2968-93-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR