2-(4-tert-Butylphenyl)ethanol 97%

Supplier: Ambeed
Warning

A109799-25G A109799-5G A109799-10G
103797-562EA 8 USD
103797-562 103823-368 77233-496 77310-178 77565-930
2-(4-tert-Butylphenyl)ethanol 97%
2-(4-tert-Butylphenyl)ethanol

Precaución: For research use only.

Formula: C₁₂H₁₈O
Melting Pt: 32…35 °C
Storage Temperature: Ambient
MDL Number: MFCD00236023
CAS Number: 5406-86-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR