2-(4-(tert-Butyl)phenyl)acetonitrile 95%

Supplier: Ambeed
Warning

Synonyms: 4-tert-Butylphenylacetonitrile, 4-tert-Butylbenzyl cyanide

A154268-100G A154268-10G A154268-500G A154268-5G A154268-25G
103842-804EA 9 USD
103842-804 76719-860 76719-862 76719-864 77300-046 77318-510
2-(4-(tert-Butyl)phenyl)acetonitrile 95%
2-(4-(tert-Butyl)phenyl)acetonitrile

Precaución: for Research Use Only

Formula: C₁₂H₁₅N
MW: 173.2500 g/mol
Flash Pt: 120.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00128112
CAS Number: 3288-99-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR