4-tert-Butylphenylacetic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-(4-(tert-Butyl)phenyl)acetic acid

A116971-100G A116971-10G A116971-250MG A116971-25G A116971-1G A116971-5G
103798-812EA 6 USD
103798-812 103824-684 76718-356 76718-358 76718-360 76718-362 77297-504 77311-476
4-tert-Butylphenylacetic acid 98%
4-tert-Butylphenylacetic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₂
MW: 192.26 g/mol
Boiling Pt: 302.8 °C (760 mm Hg)
Melting Pt: 77 °C
Flash Pt: 199.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00082593
CAS Number: 32857-63-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR