4-(Methylsulfonyl)phenylacetic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-(4-(Methylsulphonyl)phenyl)acetic acid

A314848-1G A314848-25G A314848-5G A314848-100G A314848-10G
103825-048EA 5 USD
103825-048 103835-972 76838-794 76838-796 76838-798 77251-034 77251-036
4-(Methylsulfonyl)phenylacetic acid 97%
4-(Methylsulfonyl)phenylacetic acid

Precaución: for Research Use Only

Formula: C₉H₁₀O₄S
MW: 214.24 g/mol
Melting Pt: 134…138 °C
Flash Pt: 221.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00216495
CAS Number: 90536-66-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR