2-(4-(Methylamino)phenyl)benzo[d]thiazol-6-ol 95%

Supplier: Ambeed
Warning

Synonyms: Pittsburgh Compound-B

A186835-250MG A186835-25MG A186835-1G A186835-100MG
103815-968EA 1097 USD
103815-968 76985-944 76985-946 77302-044 77611-808
2-(4-(Methylamino)phenyl)benzo[d]thiazol-6-ol 95%
2-(4-(Methylamino)phenyl)benzo[d]thiazol-6-ol

Precaución: for Research Use Only

Formula: C₁₄H₁₂N₂OS
MW: 256.32 g/mol
Boiling Pt: 475.7±51.0 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD17018727
CAS Number: 566169-93-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR