(4-(Hydroxymethyl)phenyl)acetic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-(4-(Hydroxymethyl)phenyl)acetic acid, 4-(Hydroxymethyl)phenylacetic acid

A314888-1G A314888-25G A314888-5G A314888-10G A314888-250MG A314888-100G
103825-050EA 6 USD
103825-050 103835-974 76838-800 76838-802 76838-804 77251-038 77251-040 77616-378
(4-(Hydroxymethyl)phenyl)acetic acid 97%
(4-(Hydroxymethyl)phenyl)acetic acid

Precaución: for Research Use Only

Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Melting Pt: 131…134 °C
Storage Temperature: Ambient
MDL Number: MFCD00065692
CAS Number: 73401-74-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR