Thioflavin T 95%

Supplier: Ambeed
Warning

Synonyms: CI 49005, Thioflavine T, Basic Yellow 1, 2-(4-(Dimethylamino)phenyl)-3,6-dimethylbenzo[d]thiazol-3-ium chloride, Setoflavine T

A138710-10G A138710-5G A138710-1G
103802-652EA 7 USD
103802-652 76688-922 76688-924 77299-008
Thioflavin T 95%
Thioflavin T

Precaución: for Research Use Only

Formula: C₁₇H₁₉ClN₂S
MW: 318.86 g/mol
Melting Pt: 212 °C
Storage Temperature: Ambient
MDL Number: MFCD00011944
CAS Number: 2390-54-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR