2-(4-(Difluoromethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%

Supplier: Ambeed
Danger

A187436-250MG A187436-1G A187436-5G
103816-012EA 30 USD
103816-012 103846-602 77012-172 77302-078 77302-080
2-(4-(Difluoromethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%
2-(4-(Difluoromethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₃H₁₇BF₂O₂
MW: 254.08 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16996278
CAS Number: 1234319-14-2
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR