4-Bromomethyl-3-fluorobenzeneboronic acid pinacol ester 95%

Supplier: Ambeed
Warning

Synonyms: 4-Bromomethyl-3-fluorophenylboronic acid pinacol ester, 2-(4-(Bromomethyl)-3-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1,3,2-Dioxaborolane, 2-[4-(bromomethyl)-3-fluorophenyl]-4,4,5,5-tetramethyl-, 2-[4-(Bromomethyl)-3-fluorophenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

A152680-100MG A152680-250MG A152680-1G A152680-5G
103804-418EA 25 USD
103804-418 103842-590 76871-664 76871-666 77318-216 77318-218
4-Bromomethyl-3-fluorobenzeneboronic acid pinacol ester 95%
4-Bromomethyl-3-fluorobenzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₇BBrFO₂
MW: 314.99 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11855987
CAS Number: 1150271-74-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR