(4-Benzyloxyphenyl)acetic acid 95%
Supplier: Ambeed
|
Warning
|
A114436-100G
A114436-10G
A114436-25G
A114436-1G
A114436-5G
103798-404EA
5
USD
103798-404
103824-242
76781-290
76781-292
76781-294
77297-308
77311-054
(4-Benzyloxyphenyl)acetic acid 95%
(4-Benzyloxyphenyl)acetic acid
Precaución: for Research Use Only
Formula:
(C₆H₅CH₂OC₆H₄)CH₂COOH Boiling Pt: 418.8 °C (760 mm Hg) Melting Pt: 119…121 °C Flash Pt: 158.5 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017540 CAS Number: 6547-53-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...