4-Benzyloxy-3-methoxyphenylacetonitrile 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
2-(4-(Benzyloxy)-3-methoxyphenyl)acetonitrile
A265293-10G
A265293-250MG
A265293-1G
A265293-25G
A265293-5G
103812-720EA
24
USD
103812-720
103852-614
77031-836
77031-838
77092-286
77092-288
77092-290
4-Benzyloxy-3-methoxyphenylacetonitrile 98%
4-Benzyloxy-3-methoxyphenylacetonitrile
Precaución: for Research Use Only
Formula:
C₁₆H₁₅NO₂ MW: 253.30 g/mol Boiling Pt: 410.5 °C (760 mm Hg) Flash Pt: 148.2 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016394 CAS Number: 1700-29-4 UN: 3439 ADR: 6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...