2-(4-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)phenyl)-1,1,1-trifluoropropan-2-ol 97%
Supplier: Ambeed
|
Warning
|
A101632-250MG
A101632-50MG
A101632-1G
103796-054EA
591
USD
103796-054
103821-750
77135-512
77135-514
77282-776
2-(4-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)phenyl)-1,1,1-trifluoropropan-2-ol 97%
2-(4-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)phenyl)-1,1,1-trifluoropropan-2-ol
Precaución: for Research Use Only
Formula:
C₁₄H₁₈BF₃O₃ MW: 302.1 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD28133801 CAS Number: 1467061-57-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...