4-(2-Pyridinyl)phenylboronic acid, pinacol ester 95%

Supplier: Ambeed
Warning

Synonyms: 2-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)pyridine

A106309-100MG A106309-250MG A106309-1G
103797-024EA 24 USD
103797-024 103822-776 77025-454 77025-456 77309-634
4-(2-Pyridinyl)phenylboronic acid, pinacol ester 95%
4-(2-Pyridinyl)phenylboronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₇H₂₀BNO₂
MW: 281.157 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11973624
CAS Number: 908350-80-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR