1-(2-Hydroxyethyl)-1H-pyrazole-4-boronic acid, pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: 1-Hydroxy-2-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl]ethane, 2-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl]ethan-1-ol, 2-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl)ethanol

A273098-100MG A273098-250MG
103813-128EA 18 USD
103813-128 103853-044 76816-244 77612-590
1-(2-Hydroxyethyl)-1H-pyrazole-4-boronic acid, pinacol ester 97%
1-(2-Hydroxyethyl)-1H-pyrazole-4-boronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₁H₁₉BN₂O₃
MW: 238.091 g/mol
Storage Temperature: Freezer
MDL Number: MFCD12033564
CAS Number: 1040377-08-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR