2-(4-(2,2-Difluoroethoxy)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%

Supplier: Ambeed
Warning

A127149-10G A127149-250MG A127149-1G
103800-920EA 372 USD
103800-920 76983-498 76983-500 77298-180
2-(4-(2,2-Difluoroethoxy)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%
2-(4-(2,2-Difluoroethoxy)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₄H₁₉BF₂O₃
MW: 284.11 g/mol
Boiling Pt: 346.705 °C (760 mm Hg)
Flash Pt: 163.482 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08706127
CAS Number: 864754-48-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR