4-[(2,4-Dimethoxyphenyl)-(fmoc-amino)methyl]phenoxyacetic acid 98%

Supplier: Ambeed
Warning

A218583-10G A218583-1G A218583-25G A218583-5G A218583-100G
103818-720EA 6 USD
103818-720 103849-590 76999-842 77088-116 77303-494 77303-496 77593-424
4-[(2,4-Dimethoxyphenyl)-(fmoc-amino)methyl]phenoxyacetic acid 98%
4-[(2,4-Dimethoxyphenyl)-(fmoc-amino)methyl]phenoxyacetic acid

Precaución: for Research Use Only

Formula: C₃₂H₂₉NO₇
Boiling Pt: 776.3 °C (760 mm Hg)
Melting Pt: 180 °C
Flash Pt: 423.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00153509
CAS Number: 126828-35-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR