N-Isovalerylglycine 97%

Supplier: Ambeed
Warning

Synonyms: 2-(3-Methylbutanamido)acetic acid, N-Isovaleroylglycine

A106642-250MG A106642-25G A106642-1G A106642-5G
103797-054EA 33 USD
103797-054 103822-810 76717-866 76717-868 77309-664 77309-666
N-Isovalerylglycine 97%
N-Isovalerylglycine

Precaución: for Research Use Only

Formula: C₇H₁₃NO₃
MW: 159.19 g/mol
Boiling Pt: 371.8 °C (760 mm Hg)
Melting Pt: 87…90 °C
Flash Pt: 178.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00059629
CAS Number: 16284-60-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR