2-(3-Methyl-5-(trifluoromethyl)phenyl)acetic acid 97%

Supplier: Ambeed
Warning

A470277-1G A470277-250MG
103828-018EA 115 USD
103828-018 103839-092 76843-002 76877-714
2-(3-Methyl-5-(trifluoromethyl)phenyl)acetic acid 97%
2-(3-Methyl-5-(trifluoromethyl)phenyl)acetic acid

Precaución: for Research Use Only

Formula: C₁₀H₉F₃O₂
MW: 218.172 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13194442
CAS Number: 1214352-00-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR