2-(3-Methoxyphenyl)ethanol 96%

Supplier: Ambeed
Warning

A329636-1G A329636-25G A329636-5G A329636-100G
103825-532EA 7 USD
103825-532 103836-488 76839-454 76839-456 76839-458 77277-808
2-(3-Methoxyphenyl)ethanol 96%
2-(3-Methoxyphenyl)ethanol

Precaución: for Research Use Only

Formula: C₉H₁₂O₂
Boiling Pt: 266.8 °C (760 mm Hg)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00002893
CAS Number: 5020-41-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR