(3-Hydroxy-4-methoxyphenyl)acetic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-(3-Hydroxy-4-methoxyphenyl)acetic acid

A209950-1G A209950-25G A209950-100G A209950-10G A209950-250MG A209950-5G
103817-972EA 12 USD
103817-972 103848-660 77087-022 77087-024 77249-054 77249-056 77249-058 77303-110
(3-Hydroxy-4-methoxyphenyl)acetic acid 98%
(3-Hydroxy-4-methoxyphenyl)acetic acid

Precaución: for Research Use Only

Formula: (C₆H₃(OH)(OCH₃))CH₂COOH
MW: 182.18 g/mol
Melting Pt: 127…132 °C
Storage Temperature: Ambient
MDL Number: MFCD00016829
CAS Number: 1131-94-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR